missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad2, p Ser250 Antibody (S02-0D1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19828-100ul
This item is not returnable.
View return policy
Description
Smad2 Monoclonal antibody specifically detects Smad2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Smad2 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S02-0D1 | |
| Unconjugated | |
| hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
| A synthetic phosphopeptide corresponding to residues surrounding Ser250 of human Smad2 (Uniprot # Q15796) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Metastasis, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 4087 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction