Learn More
Smad3 Antibody (378611), Alexa Fluor™ 405, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB4038V
Product Code. 30063576
  • € 607.00 / 0.10mL
Description
Smad3 Monoclonal antibody specifically detects Smad3 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Smad3
Monoclonal
Alexa Fluor 405
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
378611
Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3
E. coli-derived recombinant human Smad3, Ser2-Ala230, Accession # P84022
0.1 mL
Phospho Specific, Signal Transduction, Wnt Signaling Pathway
4088
Store at 4C in the dark.