missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad4 Antibody (SMAD/6309R) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14116JF585
This item is not returnable.
View return policy
Description
Smad4 Monoclonal antibody specifically detects Smad4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Smad4 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4 | |
| Recombinant human full-length (aa1-552) Smad4 protein (Uniprot: Q13485 ) | |
| 0.1 mL | |
| Signal Transduction, Stem Cell Signaling Pathway | |
| 4089 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SMAD/6309R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction