missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98884-50ul
This item is not returnable.
View return policy
Description
Smad9 Polyclonal antibody specifically detects Smad9 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Smad9 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:1500-1:50000 | |
| MAD homolog 9, MAD, mothers against decapentaplegic homolog 9 (Drosophila), Madh6, MADH6mothers against decapentaplegic homolog 9, MADH9Mothers against decapentaplegic, drosophila, homolog of, 9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9SMAD8B, SMAD, mothers against DPP homolog 9, SMAD, mothers against DPP homolog 9 (Drosophila), SMAD8, SMAD8A, Smad9 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Smad9. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 4093 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction