missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM106A Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41165H
This item is not returnable.
View return policy
Description
TMEM106A Polyclonal antibody specifically detects TMEM106A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMEM106A | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| Transmembrane Protein 106A | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human TMEM106A. The immunogen is located within the first 50 amino acids of TMEM106A. Amino Acid Squence: KAVNYSSTGSSKSF | |
| 0.1 mL | |
| Neuroscience | |
| 113277 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction