missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad9 Antibody (PCRP-SMAD9-2F4) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14117JF585
This item is not returnable.
View return policy
Description
Smad9 Monoclonal antibody specifically detects Smad9 in Human samples. It is validated for Flow Cytometry,Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| Smad9 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| MAD homolog 9, MAD, mothers against decapentaplegic homolog 9 (Drosophila), Madh6, MADH6mothers against decapentaplegic homolog 9, MADH9Mothers against decapentaplegic, drosophila, homolog of, 9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9SMAD8B, SMAD, mothers against DPP homolog 9, SMAD, mothers against DPP homolog 9 (Drosophila), SMAD8, SMAD8A, Smad9 | |
| Recombinant full-length human Smad9 protein (Uniprot: O15198) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-SMAD9-2F4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4093 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction