Learn More
Scientific Industries SI™ Small Ampule/Tube Attachment
Click to view available options
Holds:
4 x 15 to 17 mm dia. Tubes
Unit Size:
Each
Description
- Used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
- Tubes and ampules snap into place for vigorous end-to-end agitation
- Holds up to four 15 to 17mm diameter tubes or ampules
Specifications
Specifications
| Holds | 4 x 15 to 17 mm dia. Tubes |
| For Use With (Equipment) | Vortex-Genie 2 family of mixers, Vortex-Genie Pulse |
| No. of Positions | 4 |