Learn More
Daklapack X10000 Gripbag 30% rPE 55 x 65 mm
Brand:  Daklapack OT112RPE
Encompass_Preferred
X10000 Gripbag 30% rPE 55 x 65 mm
Product Code. 17849168
  • € 414.59 / Pack of 10000
Only null left
Estimated Shipment: 13-01-2026