Learn More
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 13072 cSt (40°C), Paragon Scientific™
Click to view available options
Quantity:
60 mL
Unit Size:
60mL
Description
- Fully traceable to international standards
- Ensures full compliance to ASTM & IP test method protocol
- Certified in strict accordance with ASTM D2162
- 60mL lower volume cost effective solution
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Specifications
Specifications
| Quantity | 60 mL |
Nominal values quoted. Certified Values will vary from batch to batch.