missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smallpox B5R Antibody (1J2F3), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24699FR
This item is not returnable.
View return policy
Description
Smallpox B5R Monoclonal specifically detects Smallpox B5R in Virus samples. It is validated for Western Blot.
Specifications
| Smallpox B5R | |
| Monoclonal | |
| DyLight 680 | |
| P33823 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1486562 | |
| Virus | |
| Purified |
| Western Blot | |
| 1J2F3 | |
| Western Blot | |
| B5R protein, B6R, B7R, B7R protein, EEV membrane glycoprotein, Glycoprotein, H7R, H7R protein, ORF5R, VARVgp175 | |
| Amino acids 33-47 (IQEYNLIRTYKKVDK) ofthe B5R protein were used as the immunogen (NCBI Reference Sequence: NP_042218.1) | |
| 0.1 mL | |
| Primary | |
| The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in Human, Chimpanzee, Dog, Rat, Mouse, Chicken and Zebrafish and 92% homologous in Xenopus. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction