missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556509
This item is not returnable.
View return policy
Description
Immunogen sequence: KAAEPTVDLL GLDGPAVAPV TNGNTTVPPL NDDLDIFGPM ISNPLPATVM PPAQGTPSAP AAATLSTVTS GDLDLFTEQT TKSEEVAKKQ LSKDSIL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
The protein encoded by this gene is similar to the mouse stromal membrane-associated protein-1. This similarity suggests that this human gene product is also a type II membrane glycoprotein involved in the erythropoietic stimulatory activity of stromal cells. Alternate splicing results in multiple transcript variants encoding different isoforms.
Specifications
| SMAP1 | |
| Polyclonal | |
| Unconjugated | |
| SMAP1 | |
| 1700056O10Rik; 4921514B13Rik; 4921525H11Rik; AI462175; small ArfGAP 1; SMAP1; SMAP-1; stromal membrane-associated GTPase-activating protein 1; stromal membrane-associated protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60682, 684800, 98366 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IYB5, Q91VZ6 | |
| SMAP1 | |
| Recombinant protein corresponding to Human SMAP1. Recombinant protein control fragment (Product #RP-94684). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction