missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SmarcAL1 Antibody (PCRP-SMARCAL1-1C4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20733FR
This item is not returnable.
View return policy
Description
SmarcAL1 Monoclonal antibody specifically detects SmarcAL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SmarcAL1 | |
| Monoclonal | |
| DyLight 680 | |
| ATP-driven annealing helicase, EC 3.6.1, EC 3.6.4.-, HARPSWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily A-like protein 1, HepA-related protein, HHARP, SIOD, SMARCA-like protein 1, subfamily a-like 1, Sucrose nonfermenting protein 2-like 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin | |
| Recombinant full-length human SmarcAL1 protein | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 50485 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin) | |
| PCRP-SMARCAL1-1C4 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction