missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCB1, Mouse anti-Human, Clone: 3E10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006598-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: YTTLATSVTLLKASEVEEILDGNDEKYKAVSISTEPPTYLREQKAKRNSQWVPTLPNSSHHLDAVPCSTTINRNRMGRDKKRTFPLCFDDHDPAVIHENASpecifications
| SMARCB1 | |
| Monoclonal | |
| Unconjugated | |
| SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 | |
| BAF47/INI1/RDT/SNF5/SNF5L1/Sfh1p/Snr1/hSNFS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3E10 | |
| In 1x PBS, pH 7.4 | |
| NM_003073 | |
| SMARCB1 | |
| SMARCB1 (NP_003064, 81 a.a. ∼ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6598 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction