missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | SMARCC1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616225
|
Novus Biologicals
NBP2-98777-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18679715
|
Novus Biologicals
NBP2-98777-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMARCC1 Polyclonal antibody specifically detects SMARCC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| SMARCC1 | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Modifiers | |
| 0.2 um filtered solution in PBS | |
| 6599 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SMARCC1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title