missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCC1 Antibody (PCRP-SMARCC1-1F1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08611F
This item is not returnable.
View return policy
Description
SMARCC1 Monoclonal antibody specifically detects SMARCC1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| SMARCC1 | |
| Monoclonal | |
| FITC | |
| BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3 | |
| Recombinant full-length human SMARCC1 protein (Uniprot: Q92922) | |
| 100 μg | |
| Chromatin Modifiers | |
| 6599 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, Immunofluorescence, Protein Array | |
| PCRP-SMARCC1-1F1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction