missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCC1 Mouse anti-Human, Clone: PCRP-SMARCC1-1F1, Novus Biologicals™
Mouse Monoclonal Antibody
€ 484.00
Specifications
| Antigen | SMARCC1 |
|---|---|
| Clone | PCRP-SMARCC1-1F1 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
SMARCC1 Monoclonal specifically detects SMARCC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| SMARCC1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| BAF155SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily C member 1, BRG1-associated factor 155, chromatin remodeling complex BAF155 subunit, CRACC1, mammalian chromatin remodeling complex BRG1-associated factor 155, Rsc8, SRG3, subfamily c, member 1, SWI/SNF complex 155 kDa subunit, SWI/SNF complex subunit SMARCC1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI3 | |
| Recombinant full-length human SMARCC1 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-SMARCC1-1F1 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 6599 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title