missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98471B
This item is not returnable.
View return policy
Description
SMARCD1 Polyclonal specifically detects SMARCD1 in Human samples. It is validated for Western Blot.
Specifications
| SMARCD1 | |
| Polyclonal | |
| Western Blot | |
| BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A, BRG1-associated factor 60A, chromatin remodeling complex BAF60A subunit, CRACD1, Rsc6p, subfamily d, member 1, SWI/SNF complex 60 kDa subunit, SWI/SNF complex 60 kDa subunit A, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 1,60 kDa BRG-1/Brm-associated factor subunit A, Swp73-like protein | |
| Produced in rabbits immunized with E. coli-derived Human SMARCD1 fragment. | |
| 0.1 mL | |
| Chromatin Research | |
| 6602 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction