missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCD1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79836DL594
This item is not returnable.
View return policy
Description
SMARCD1 Polyclonal antibody specifically detects SMARCD1 in Human samples. It is validated for Western Blot
Specifications
| SMARCD1 | |
| Polyclonal | |
| Western Blot | |
| BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A, BRG1-associated factor 60A, chromatin remodeling complex BAF60A subunit, CRACD1, Rsc6p, subfamily d, member 1, SWI/SNF complex 60 kDa subunit, SWI/SNF complex 60 kDa subunit A, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 1,60 kDa BRG-1/Brm-associated factor subunit A, Swp73-like protein | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1 using the numbering given in entry NP_003067.2 (GeneID 6602). | |
| 0.1 mL | |
| Chromatin Research | |
| 6602 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto