missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SMARCD1 Antibody (SR2544), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27807
This item is not returnable.
View return policy
Description
SMARCD1 Monoclonal antibody specifically detects SMARCD1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SMARCD1 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| SR2544 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:20 | |
| BAF60Amammalian chromatin remodeling complex BRG1-associated factor 60A, BRG1-associated factor 60A, chromatin remodeling complex BAF60A subunit, CRACD1, Rsc6p, subfamily d, member 1, SWI/SNF complex 60 kDa subunit, SWI/SNF complex 60 kDa subunit A, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily D member 1,60 kDa BRG-1/Brm-associated factor subunit A, Swp73-like protein | |
| A synthetic peptide of human SMARCD1 (Uniprot #: Q96GM5) | |
| 100 μL | |
| Chromatin Research | |
| 6602 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction