missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMARCD2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17168323
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17168323 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17168323 Supplier Invitrogen™ Supplier No. PA5110398

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RDFMLSFSTD PQDFIQEWLR SQRRDLKIIT DVIGNPEEER RAAFYHQPWA QEAVGRHIFA KVQQRRQELE QVLGIRLT.

SMARCD2 is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMARCD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SMARCD2
Gene Accession No. O54772, Q92925, Q99JR8
Gene Alias 60 kDa BRG-1/Brm-associated factor subunit B; AW322457; Baf60b; BRG1-associated factor 60B; chromatin remodeling complex BAF60B subunit; CRACD2; mammalian chromatin remodeling complex BRG1-associated factor 60B; PRO2451; Rsc6p; SMARCD2; SWI/SNF complex 60 kDa subunit B; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 2; Swp73-like protein
Gene Symbols SMARCD2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 454-531 of human SMARCD2 (NP_001091896.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6603, 83796, 83833
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.