Learn More
SMARCD3 Monoclonal Antibody (OTI4G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA811107
Encompass_Preferred
 View more versions of this product
Product Code. 17584764
  • € 540.87 / 100µL
Description
BAF60C is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined.
Specifications
Specifications
SMARCD3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6STE5
SMARCD3
Full length human recombit protein of human SMARCD3 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI4G3
Unconjugated
SMARCD3
BAF60C, CRACD3, Rsc6p
Mouse
Affinity Chromatography
RUO
6604
-20° C, Avoid Freeze/Thaw Cycles