missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Smart-Tracker™ Wireless Data Module Accessories

Maximize Thermo Scientific™ Smart-Tracker™ with additional accessories or personalize your Smart-Tracker with a series of colored bands.

Merk:  Thermo Scientific™ ST202-200

Overige gegevens : Gewicht : 0.20000kg

 Bekijk meerdere types van dit product via de

Produktcode. 15448300

  • € 99.90 / 1 stuk


Geschatte verzenddatum
bij leverancier 15-07-2019
Toevoegen aan winkelmandje

Specificatie

Specificatie

Smart-Tracker module
Temperature sensor -200/+200°C
Smart-Tracker™ Module
0.2kg
Warranty varies by country, please contact us for details
Sensor
0.24 x 0.63 x 1.02 in. (6 x 16 x 26mm) [Shipping Dimensions]
Video's