Learn More
Thermo Scientific™ Smart-Vue™ Receivers
Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.
Brand:  Thermo Scientific™ SV104-502-LSB
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11834673
  • € 889.00 / Each
Estimated Shipment
27-05-2021
Add to basket
Description
Description
Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.
Compatible With:
  • Refrigerators
  • Freezers
  • LN2 Storage
  • CO2 Incubators
  • Clean Rooms
  • Storage Areas
  • Ovens
Ordering Information:
Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.
Specifications
Specifications
Smart-Vue™ Wireless Monitoring Solutions
TCP/IP, RJ45, 25/500mW Receiver w/universal power supply and client server software
Universal (100/240V) power supply, LAN CAT 5 cable, wall mounting holder with Velcro, CD-ROM with Smart-Vue client software, software drivers and user manuals
Receiver, 868MHz
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance. Meets UL, CUL, CE, and CSA standards for safety and performance.
Cold Storage
868MHz