missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Smart-Vue™ Event Notification Hardware

Receive immediate notification by means of your preferred alert method, including audible/visual siren, fax, telephone, email or text messaging.

Brand:  Thermo Scientific™ SV509-500-LSB

Additional Details : Weight : 0.33120kg Transport : UN number : 3091 Chem class : 9

 View more versions of this product

Product Code. 11894723

  • € 484.00 / Each


Estimated Shipment: 03-08-2021
Add to basket

Description

Description

Receive immediate notification by means of your preferred alert method, including audible/visual siren, fax, telephone, email or text messaging. Thermo Scientific™ Smart-Vue™ Event Notification Hardware delivers the continuous monitoring, secure data logging and automatic audit trails required to comply with SOP, cGxP and 21 CFR part 11 regulatory standards.

Local RF Audible and Visual Alarm

  • Universal (100V.240V) to 12 volt power supply
  • 3.6V battery
  • Velcro
  • Hardware
  • Customer reference letter

Voice and Fax Alert

  • USB 1.0 or 2.0 powered
  • Fits standard RJ11 telephone line

Ordering Information:

Availability of Smart-Vue products vary by country. Please ask your local Sales Representative to evaluate a Smart-Vue wireless monitoring solution that best meets your specific requirements. Specifications, terms and pricing are subject to change. Not all products are available in all countries.

Specifications

Specifications

Standard RJ11 Telephone Line
Smart-Vue Voice and Fax Alert
USB 1.0 or 2.0-powered voice/fax alert
915 MHz
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Cold Storage
Voice and Fax Alert
Voice and Fax Alert
Videos
SDS
Documents
Certificates