Learn More
Tork Light Duty Cleaning Cloths W4
Shop All Tork Products
Click to view available options
Color:
White
Unit Size:
Pack of 4
Description
- Multipurpose cleaning cloths for light-duty cleaning tasks.
- Use these durable cloths wet or dry to clean dust and dirt, and you can rinse and reuse them as needed.
- These disposable cleaning cloths are compatible with most chemical solvents, so no need to change your cleaning products.
- Compatible Top-pak dispenser has one-at-a-time dispensing to help optimise consumption and minimise waste.
Specifications
Specifications
| Product Type | Cleaning Cloth |
| Length (English) | 16.34 in. |
| Length (Metric) | 41.5 cm |
| Width (English) | 13.98 in. |
| Width (Metric) | 35.5 cm |
| Color | White |
| Disposable | Reusable |
| Packaging Quantity | 480/Cs. |
| Ply | 1-ply |