Learn More
SMC1A Monoclonal Antibody (OTI5A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809793
Encompass_Preferred
 View more versions of this product
Product Code. 17562364
  • € 540.87 / 100µL
Description
Structural Maintece of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregatior defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.
Specifications
Specifications
SMC1A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14683, Q9CU62
Smc1a
Human recombit protein fragment corresponding to amino acids 889-1016 of human SMC1A produced in E.coli.
100 μL
Primary
Human, Mouse
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI5A1
Unconjugated
SMC1A
CDLS2, DXS423E, SB1.8, SMC1, SMC1L1, SMC1alpha, SMCB
Mouse
Affinity Chromatography
RUO
24061, 8243
-20° C, Avoid Freeze/Thaw Cycles