missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-371AF647
This item is not returnable.
View return policy
Description
SMC2 Polyclonal antibody specifically detects SMC2 in Human samples. It is validated for Western Blot
Specifications
| SMC2 | |
| Polyclonal | |
| Western Blot | |
| CAP-E, CAPESMC2 structural maintenance of chromosomes 2-like 1 (yeast), Chromosome-associated protein E, FLJ10093, hCAP-EXCAP-E homolog, SMC protein 2, SMC-2, SMC2 (structural maintenance of chromosomes 2, yeast)-like 1, SMC2 structural maintenance of chromosomes 2-like 1, SMC2L1, structural maintenance of chromosomes (SMC) family member, structural maintenance of chromosomes 2, structural maintenance of chromosomes protein 2 | |
| The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human Structural Maintenance of Chromosomes 2 using the numbering given in entry NP_006435.1 (GeneID 10592). | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 10592 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction