Learn More
SMC2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-373APC
Product Code. 18592821
  • € 661.00 / 0.10mL
Description
SMC2 Polyclonal antibody specifically detects SMC2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown