Learn More
SMC2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-373G
Product Code. 18008483
  • € 686.00 / 100µL
Description
SMC2 Polyclonal specifically detects SMC2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SMC2
Polyclonal
Western Blot, Immunocytochemistry/Immunofluorescence, KnockDown Validated
CAP-E, CAPESMC2 structural maintenance of chromosomes 2-like 1 (yeast), Chromosome-associated protein E, FLJ10093, hCAP-EXCAP-E homolog, SMC protein 2, SMC-2, SMC2 (structural maintenance of chromosomes 2, yeast)-like 1, SMC2 structural maintenance of chromosomes 2-like 1, SMC2L1, structural maintenance of chromosomes (SMC) family member, structural maintenance of chromosomes 2, structural maintenance of chromosomes protein 2
Rabbit
Affinity Purified
RUO
Primary
Human
IgG
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
SMC2
The immunogen recognized by this antibody maps to a region between residue 1150 and the C-terminus (residue 1197) of human Structural Maintenance of Chromosomes 2 using the numbering given in entry NP_006435.1 (GeneID 10592).
100 μL
Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
10592
Store at 4C in the dark.