missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550767
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SMC2 protein.
CAP-E and CAP-C (Chromosome Associated Protein-E & C) are also SMC (Structural Maintenance of Chromosome) family members that form a heterodimeric complex required for mitotic chromosome condensation to achieve proper segregation of genetic information during subsequent cell division. hCAP-C/hCAP-E is a component of a multiprotein complex called condensin that interacts with phosphorylation of Histone H3 resulting in the mitotic chromosome condensation. The distribution patterns of the two heterodimeric complexes in interphase nucleus indicate independent behavior of the two complexes during cell cycle. These results suggest that the two distinct complexes are involved in different aspects of mitotic chromosome organization in humar cells.
Specifications
| SMC2 | |
| Polyclonal | |
| Unconjugated | |
| SMC2 | |
| 5730502P04Rik; AI255214; AW545314; CAP E; CAPE; CAP-E; Chromosome-associated protein E; FGF-inducible protein 16; fibroblast growth factor inducible 16; Fin16; FLJ10093; hCAP-E; PRO0324; SMC protein 2; Smc2; SMC-2; SMC2 (structural maintenance of chromosomes 2, yeast)-like 1; SMC2 protein; SMC2 structural maintenance of chromosomes 2-like 1; Smc2l1; structural maintenance of chromosomes (SMC) family member, chromosome-associated protein E; structural maintenance of chromosomes 2; structural maintenance of chromosomes protein 2; XCAP-E homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10592 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 2.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O95347 | |
| SMC2 | |
| Synthetic peptide corresponding to a region derived from internal residues of human structural maintenance of chromosomes 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction