missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC3, p Ser103 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37694B
This item is not returnable.
View return policy
Description
SMC3 Polyclonal antibody specifically detects SMC3 in Human samples. It is validated for Western Blot
Specifications
| SMC3 | |
| Polyclonal | |
| Western Blot | |
| Bamacan, BAMSMC protein 3, Basement membrane-associated chondroitin proteoglycan, BMH, Chondroitin sulfate proteoglycan 6, chondroitin sulfate proteoglycan 6 (bamacan), Chromosome-associated polypeptide, CSPG6structural maintenance of chromosomes protein 3, hCAP, HCAPbamacan, SMC-3, SMC3L1CDLS3, structural maintenance of chromosomes 3 | |
| This antibody maps to a region between residue This antibody maps to a region surrounding residue 1083 when Serine 1083 is phosphorylated of human Structural Maintenance of Chromosomes 3 using the numbering given in SwissProt entry Q9UQE7 (GeneID 9126). of human Structural Maintenance of Chromosome 3 using the numbering given in entry NP_005436.1 (GeneID 9126). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Phospho Specific | |
| 9126 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction