missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMC4 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-374B
This item is not returnable.
View return policy
Description
SMC4 Polyclonal antibody specifically detects SMC4 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| SMC4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CAP-C, CAPCSMC4 structural maintenance of chromosomes 4-like 1 (yeast), Chromosome-associated polypeptide C, hCAP-CSMC4 (structural maintenance of chromosomes 4, yeast)-like 1, SMC protein 4, SMC-4, SMC4 structural maintenance of chromosomes 4-like 1, SMC4L1, structural maintenance of chromosomes 4, structural maintenance of chromosomes protein 4, XCAP-C homolog | |
| The immunogen recognized by this antibody maps to a region between residues 600 and 650 of human Structural Maintenance of Chromosomes 4 using the numbering given in entry NP_005487.2 (GeneID 10051). | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 10051 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction