missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141402
This item is not returnable.
View return policy
Description
Suggested positive control: Ramos cell lysate. Predicted to react with Mouse, Rat, Xenopus and Chicken.
The proteins belonging to structural maintenance of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.
Specifications
| SMC4 | |
| Polyclonal | |
| Unconjugated | |
| Smc4 | |
| 2500002A22Rik; C79747; Capc; CAP-C; chromosome-associated polypeptide C; hCAP-C; SMC protein 4; SMC4; SMC-4; SMC4 structural maintenance of chromosomes 4-like 1; SMC4L1; structural maintenance of chromosomes 4; structural maintenance of chromosomes protein 4; XCAP-C homolog | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10051 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.2% gelatin and 0.05% sodium azide | |
| Q9NTJ3 | |
| Smc4 | |
| A synthetic peptide corresponding to amino acids 281-297 of human CAP-C. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction