missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCO1 Antibody (OTI1H6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74238B
This item is not returnable.
View return policy
Description
SMCO1 Monoclonal specifically detects SMCO1 in Human samples. It is validated for Western Blot.
Specifications
| SMCO1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| SMCO1 | |
| Full length recombinant protein of human SMCO1 (NP_001071125) produced in E.coli. | |
| 0.1 mL | |
| 255798.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1H6 | |
| Western Blot | |
| C3orf43, Chromosome 3 Open Reading Frame 43, Single-Pass Membrane And Coiled-Coil Domain-Containing Protein 1, Single-Pass Membrane Protein With Coiled-Coil Domains 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction