missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMCP Monoclonal Antibody (5C10A2)
GREENER_CHOICE

Product Code. 16325344
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16325344 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16325344 Supplier Invitrogen™ Supplier No. MA531913

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-31913 has been tested in indirect ELISA.

Involved in sperm motility. Its absence is associated with genetic background dependent male infertility. Infertility may be due to reduced sperm motility in the female reproductive tract and inability to penetrate the oocyte zona pellucida.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5C10A2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SMCP
Gene Accession No. P49901
Gene Alias HSMCSGEN1; MCS; MCSP; mitochondrial capsule selenoprotein; SMCP; sperm mitochondria associated cysteine rich protein; sperm mitochondria-associated cysteine-rich protein; sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161
Gene Symbols SMCP
Host Species Mouse
Immunogen Purified recombinant fragment of human SMCP (AA: 1-116) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.