missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMCP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140292
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TQDKGCQTQQ QPHSPQNESR PS Highest antigen sequence indentity to the following orthologs: Rat - 45%.
Involved in sperm motility. Its absence is associated with genetic background dependent male infertility. Infertility may be due to reduced sperm motility in the female reproductive tract and inability to penetrate the oocyte zona pellucida.
Specifications
| SMCP | |
| Polyclonal | |
| Unconjugated | |
| SMCP | |
| HSMCSGEN1; MCS; MCSP; mitochondrial capsule selenoprotein; SMCP; sperm mitochondria associated cysteine rich protein; sperm mitochondria-associated cysteine-rich protein; sperm mitochondrial-associated cysteine-rich protein; testicular tissue protein Li 119; testicular tissue protein Li 161 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4184 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49901 | |
| SMCP | |
| Recombinant protein corresponding to Human SMCP. Recombinant protein control fragment (Product #RP-109613). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction