missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMCR7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16604235
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16604235 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16604235 Supplier Invitrogen™ Supplier No. PA599984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SMCR7/MID49.

This gene encodes an outer mitochondrial membrane protein that functions in the regulation of mitochondrial morphology. It can directly recruit the fission mediator dynamin-related protein 1 (Drp1) to the mitochondrial surface. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCR7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MIEF2
Gene Accession No. Q5NCS9, Q96C03
Gene Alias AI482195; Gm11; Mid49; MIEF2; mitochondrial dynamic protein MID49; mitochondrial dynamic protein of 49 kDa; mitochondrial dynamic protein of 49 kDa homolog; mitochondrial dynamics protein MID49; Mitochondrial dynamics protein of 49 kDa; mitochondrial dynamics protein of 49 kDa homolog; Mitochondrial elongation factor 2; RGD1560728; SMCR7; Smith-Magenis syndrome chromosomal region candidate gene 7 protein; smith-Magenis syndrome chromosomal region candidate gene 7 protein homolog; Smith-Magenis syndrome chromosome region, candidate 7; Smith-Magenis syndrome chromosome region, candidate 7 homolog
Gene Symbols MIEF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MIEF2(Accession Q96C03), corresponding to amino acid residues L325-R375.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125170, 237781, 497916
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.