missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMCX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20422
This item is not returnable.
View return policy
Description
SMCX Polyclonal specifically detects SMCX in Human samples. It is validated for Western Blot.
Specifications
| SMCX | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| DXS1272EMRXJ, EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1C, JARID1Clysine-specific demethylase 5C, jumonji, AT rich interactive domain 1C, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, lysine (K)-specific demethylase 5C, MRXSJ, Protein SmcX, Protein Xe169, Smcx homolog, X chromosome, SMCXselected cDNA on X, Smcy homolog, X-linked, Smcy homolog, X-linked (mouse), XE169JmjC domain-containing protein SMCX | |
| Rabbit | |
| 176 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Dog (99%), Bovine (99%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000 | |
| P41229 | |
| KDM5C | |
| Recombinant protein encompassing a sequence within the N-terminus region of human JARID1C. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 8242 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction