missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SMCX Antibody (PCRP-KDM5C-1A11) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27035H
This item is not returnable.
View return policy
Description
SMCX Monoclonal antibody specifically detects SMCX in Human samples. It is validated for Flow Cytometry
Specifications
| SMCX | |
| Monoclonal | |
| HRP | |
| DXS1272EMRXJ, EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1C, JARID1Clysine-specific demethylase 5C, jumonji, AT rich interactive domain 1C, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, lysine (K)-specific demethylase 5C, MRXSJ, Protein SmcX, Protein Xe169, Smcx homolog, X chromosome, SMCXselected cDNA on X, Smcy homolog, X-linked, Smcy homolog, X-linked (mouse), XE169JmjC domain-containing protein SMCX | |
| Recombinant full-length human SMCX protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-KDM5C-1A11 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 8242 | |
| Store at 4°C in the dark. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction