missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SMDT1 Antibody [mFluor Violet 500 SE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-26913MFV500
This item is not returnable.
View return policy
Description
SMDT1 Polyclonal antibody specifically detects SMDT1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| SMDT1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 91689 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| chromosome 22 open reading frame 32, DDDD, dJ186O1.1, hypothetical protein LOC91689 | |
| Produced in rabbits immunized with E. coli-derived Human SMDT1 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction