missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smek1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94709-0.02ml
This item is not returnable.
View return policy
Description
Smek1 Polyclonal antibody specifically detects Smek1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Smek1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| FLFL1, FLJ20707, KIAA2010MSTP033, PP4R3A, PPP4R3A, serine/threonine-protein phosphatase 4 regulatory subunit 3A, SMEK homolog 1, SMEK homolog 1, suppressor of mek1 (Dictyostelium), smk1, smk-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 395-594 of human SMEK1 (NP_001271210.1).VQTFKGLKLRFEQQRERQDNPKLDSMRSILRNHRYRRDARTLEDEEEMWFNTDEDDMEDGEAVVSPSDKTKNDDDIMDPISKFMERKKLKESEEKEVLLKTNLSGRQSPSFKLSLSSGTKTNLTSQSSTTNLPGSPGSPGSPGSPGSPGSVPKNTSQTAAITTKGGLVGLVDYPDDDEDDDEDEDKEDTLPLSKKAKFDS | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55671 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction