missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smek1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2879
This item is not returnable.
View return policy
Description
Smek1 Polyclonal specifically detects Smek1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Smek1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| FLFL1, FLJ20707, KIAA2010MSTP033, PP4R3A, PPP4R3A, serine/threonine-protein phosphatase 4 regulatory subunit 3A, SMEK homolog 1, SMEK homolog 1, suppressor of mek1 (Dictyostelium), smk1, smk-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55671 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q6IN85 | |
| PPP4R3A | |
| The immunogen recognized by this antibody maps to a region between residue 770 and the C-terminus (residue 820) of human Protein Phosphatase 4, Regulatory Subunit 3 Alpha (KIAA2010) using the numbering given in entry NP_115949.3 | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey,Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheekedgibbon. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction