missing translation for 'onlineSavingsMsg'
Learn More

SMER18, MedChemExpress

Product Code. 16467506
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467506 100 mg 100mg
16497568 10 mg 10mg
16460048 5 mg 5mg
16403597 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467506 Supplier Medchem Express Supplier No. HY18672

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SMER18 is a small molecule enhancer of rapamycin which act as a mTOR-independent autophagy inducer.

SMER18 is a small molecule enhancer of rapamycin which act as a mTOR-independent autophagy inducer.

Specifications

Chemical Name or Material SMER18
CAS 944153-47-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H14ClNO2
Quantity 100 mg
SMILES C/C(NC1=CC=CC(O)=C1)=C/C(C2=CC=C(Cl)C=C2)=O
Molecular Weight (g/mol) 287.74
Formula Weight 287.74
Percent Purity 98.6%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.