missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMG7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20423
This item is not returnable.
View return policy
Description
SMG7 Polyclonal specifically detects SMG7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SMG7 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| SMG7 | |
| Recombinant protein encompassing a sequence within the center region of human SMG7. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 9887 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| breast cancer-associated antigen SGA-56M, C1orf16, chromosome 1 open reading frame 16, EST1 telomerase component homolog C, EST1CKIAA0250SGA56M, EST1-like protein C, ever shorter telomeres 1C, FLJ23717, hSMG-7, protein SMG7, SMG-7, SMG-7 homolog, Smg-7 homolog, nonsense mediated mRNA decay factor (C. elegans) | |
| Rabbit | |
| 132 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction