missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMG7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17188323
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17188323 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17188323 Supplier Invitrogen™ Supplier No. PA5110400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMG7 Polyclonal Antibody for ELISA, Western Blot

SMG7 is involved in nonsense-mediated mRNA decay (Fukuhara et al., 2005 [PubMed 15721257]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMG7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SMG7
Gene Accession No. Q5RJH6, Q92540
Gene Alias 9430023P16Rik; breast cancer-associated antigen SGA-56M; C1orf16; EST1 telomerase component homolog C; Est1c; EST1-like protein C; ever shorter telomeres 1C; hSMG-7; KIAA0250; mKIAA0250; Protein SMG7; SGA56M; Smg7; SMG-7 homolog; smg-7 homolog, nonsense mediated mRNA decay factor; Smg-7 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG7 nonsense mediated mRNA decay factor; SMG7, nonsense mediated mRNA decay factor
Gene Symbols SMG7
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226517, 360855, 9887
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.