missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMIM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 589.00
Specifications
| Antigen | SMIM1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623946
|
Novus Biologicals
NBP2-38120-25ul |
25 μL |
€ 216.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18171529
|
Novus Biologicals
NBP2-38120 |
0.1 mL |
€ 589.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMIM1 Polyclonal specifically detects SMIM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SMIM1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Small Integral Membrane Protein 1, Small Integral Membrane Protein 1 (Vel Blood Group), Vel, Vel Blood Group Antigen | |
| SMIM1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| B2RUZ4 | |
| 388588 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QPQESHVHYSRWEDGSRDGVSLGAVSSTEEASRCRRISQRLCTGKL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title