Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Culture Holders and Transfer Devices
Product Code. 15795068
Encompass
Click to view available options
Includes:
Female Luer
Unit Size:
Pack of 50
Product Code. 15795068
Brand: ICU Medical 96005
This item requires a price update prior to being ordered.
Request a Price for this item here.
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
  • Eliminate potential exposures and holder reuse
  • Provide safe needleless transfer after a syringe draw
Specifications
For Use With (Application) Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw
Includes Female Luer
Product Type Blood Culture Device
Model Saf-T Holder