missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN Antibody (2B1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-1936AF700
This item is not returnable.
View return policy
Description
SMN Monoclonal specifically detects SMN in Human, Mouse, Rat, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SMN | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Q16637 | |
| SMN1 | |
| Purified recombinant His6-tagged human SMN protein. [Swiss-Prot# Q16637] | |
| 0.1 mL | |
| Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators | |
| 6606 | |
| Human, Mouse, Rat, Primate, Xenopus species | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2B1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, mouse, rat, monkey and Xenopus. Other species not tested. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction