missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN Antibody (8B10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26312-50ul
This item is not returnable.
View return policy
Description
SMN Monoclonal antibody specifically detects SMN in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| SMN | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 8B10 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric | |
| A synthesized peptide derived from Human SMN [UniProt Q16637] | |
| 50 μL | |
| Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators | |
| 6606 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction