Learn More
SMN Antibody (SMN1/1596), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-45072
Product Code. 30448880
  • € 593.00 / 100µg
Description
SMN Monoclonal antibody specifically detects SMN in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SMN
Monoclonal
1 mg/mL
Immunohistochemistry : 1-2ug/mL
Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric
Recombinant full-length human SMN protein (Uniprot: Q16637).
100 μg
Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators
6606
Store at -20 to -84°C. Avoid freeze-thaw cycles.
Purified
Immunohistochemistry
SMN1/1596
Unconjugated
10 mM PBS
Mouse
Protein G purified